Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020405.4 | NP_065138.2 | 13 | 1576 | ||
GenBank mRNA | AF279144.2 | - | 13 | 1446 | ||
AF378753.1 | - | 13 | 1446 | |||
AK093589.1 | - | 12 | 1875 | |||
AK127539.1 | - | 12 | 1757 | |||
AK303699.1 | - | 12 | 1263 | |||
AK312999.1 | - | 13 | 1398 | |||
AL833517.1 | - | 5 | 452 | |||
AY704670.1 | - | 13 | 1810 | |||
AY704671.1 | - | 14 | 2010 | |||
AY704672.1 | - | 13 | 1756 | |||
BC036059.1 | - | 13 | 1542 | 5298499 |