Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018226.4 | NP_060696.4 | 10 | 1788 | ||
XM_005247036.4 | XP_005247093.1 | 10 | 1816 | |||
GenBank mRNA | AF300795.1 | - | 10 | 1494 | ||
AK001668.1 | - | 6 | 752 | |||
AK130185.1 | - | 3 | 356 | |||
AK130805.1 | - | 3 | 2722 | |||
AL512754.1 | - | 3 | 373 | |||
BC017301.2 | - | 7 | 940 | |||
BC067258.1 | - | 10 | 1363 | 6183046 | ||
BC082975.1 | - | 10 | 1688 | 6379537 | ||
BC099906.1 | - | 8 | 1901 | 5311883 |