Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199951.2 | NP_001186880.1 | 3, 4 | 536 | ||
NM_020426.3 | NP_065159.1 | 3, 4 | 650 | |||
GenBank mRNA | AY359018.1 | - | 2, 3 | 454 | ||
AY742214.1 | - | 2, 3 | 426 | |||
BC054481.1 | - | 3, 4 | 496 | 6620264 | ||
BG195727.1 | - | 2, 3 | 504 | |||
GQ472214.1 | - | 3, 4 | 496 |