Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 3 | 464 | ||
NM_001079685.2 | NP_001073153.1 | 3 | 443 | |||
NM_001287336.1 | NP_001274265.1 | 2 | 356 | |||
NM_020444.4 | NP_065177.2 | 4 | 551 | |||
XM_005265941.1 | XP_005265998.1 | 3 | 364 | |||
XM_005265946.4 | XP_005266003.1 | 3 | 459 | |||
GenBank mRNA | AK315428.1 | - | 4 | 508 | ||
AL390217.1 | - | 3 | 422 | |||
BC010448.1 | - | 4 | 486 | 4155679 | ||
BC017097.1 | - | 4 | 508 | 3529175 | ||
BX648859.1 | - | 4 | 550 | |||
CR457331.1 | - | 2 | 178 | |||
DA789543.1 | - | 3 | 433 | |||
DQ073392.1 | - | 2 | 178 | |||
HY119081.1 | - | 2 | 356 |