Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005855.1 | NP_001005855.1 | 10 | 855 | ||
NM_020452.3 | NP_065185.1 | 10 | 774 | |||
XM_005245355.2 | XP_005245412.1 | 9 | 921 | |||
XM_005245356.3 | XP_005245413.1 | 10 | 874 | |||
XM_017001871.1 | XP_016857360.1 | 9 | 921 | |||
XM_017001872.1 | XP_016857361.1 | 10 | 870 | |||
GenBank mRNA | AK054886.1 | - | 9 | 921 | ||
AY302537.1 | - | 10 | 774 | |||
BC069264.1 | - | 10 | 862 |