Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 4 | 995 | ||
NM_001288953.1 | NP_001275882.1 | 5 | 986 | |||
NM_001288955.1 | NP_001275884.1 | 4 | 995 | |||
NM_020458.3 | NP_065191.2 | 4 | 995 | |||
XM_005264439.3 | XP_005264496.1 | 5 | 1148 | |||
XM_011532998.2 | XP_011531300.1 | 6 | 1213 | |||
XM_011532999.2 | XP_011531301.1 | 4 | 938 | |||
XM_017004524.1 | XP_016860013.1 | 4 | 939 | |||
XM_017004525.1 | XP_016860014.1 | 4 | 480 | |||
XM_017004526.1 | XP_016860015.1 | 4 | 939 | |||
XM_017004527.1 | XP_016860016.1 | 4 | 1090 | |||
XM_017004528.1 | XP_016860017.1 | 5 | 1155 | |||
XM_017004529.1 | XP_016860018.1 | 4 | 938 | |||
XM_017004530.1 | XP_016860019.1 | 4 | 939 | |||
GenBank mRNA | AB032966.2 | - | 5 | 575 | ||
AK056464.1 | - | 5 | 958 | |||
AL834383.1 | - | 4 | 995 | |||
BC065554.1 | - | 1 | 113 | |||
BC111487.1 | - | 4 | 883 | 6041916 | ||
BC114365.2 | - | 4 | 626 | 40038218 |