Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122964.2 | NP_001116436.2 | 8 | 1764 | ||
NM_001282850.1 | NP_001269779.1 | 8 | 1764 | |||
NM_020463.3 | NP_065196.1 | 8 | 1764 | |||
XM_005264442.2 | XP_005264499.1 | 8 | 1764 | |||
XM_005264444.2 | XP_005264501.1 | 8 | 1764 | |||
XM_005264445.2 | XP_005264502.1 | 8 | 1764 | |||
XM_005264446.2 | XP_005264503.1 | 8 | 1764 | |||
XM_005264447.2 | XP_005264504.1 | 7 | 1140 | |||
XM_017004531.1 | XP_016860020.1 | 8 | 1764 | |||
XM_017004532.1 | XP_016860021.1 | 7 | 1140 | |||
XM_017004533.1 | XP_016860022.1 | 7 | 1140 | |||
XM_017004534.1 | XP_016860023.1 | 7 | 1140 | |||
XM_017004535.1 | XP_016860024.1 | 7 | 1140 | |||
XM_017004536.1 | XP_016860025.1 | 7 | 1140 | |||
XM_017004537.1 | XP_016860026.1 | 9 | 1955 | |||
XM_017004538.1 | XP_016860027.1 | 9 | 1867 | |||
XM_017004539.1 | XP_016860028.1 | 9 | 1847 | |||
XM_017004540.1 | XP_016860029.1 | 9 | 1867 | |||
GenBank mRNA | AB037808.1 | - | 8 | 1627 | ||
AK026437.1 | - | 5 | 1042 | |||
AK056036.1 | - | 8 | 1661 | |||
AY825268.1 | - | 8 | 1324 | |||
AY825269.1 | - | 8 | 1324 | |||
BC006215.2 | - | 8 | 1635 | 3140319 | ||
BC060855.1 | - | 8 | 1653 | 5259789 |