Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A10 SIRNA Assays ›
Gene Symbol:
SLC4A10
Gene Name:
solute carrier family 4 member 10
Gene Aliases:
NBCn2, NCBE
Chromosome Location:
Chr.2: 161624335 - 161985276 on Build GRCh38
UniGene ID:
Hs.333958
Species:
Human
siRNA ID
HSS126181
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178015.1
NP_001171486.1
12
1600
NM_001178016.1
NP_001171487.1
12
1646
NM_022058.3
NP_071341.2
11
1510
XM_005246694.4
XP_005246751.1
13
1655
XM_005246695.4
XP_005246752.1
12
1565
XM_011511508.2
XP_011509810.1
12
1443
XM_011511509.2
XP_011509811.1
12
1440
XM_011511510.2
XP_011509812.1
12
1443
XM_011511511.2
XP_011509813.1
11
1353
XM_011511512.2
XP_011509814.1
11
1350
XM_011511513.2
XP_011509815.1
15
1932
XM_017004541.1
XP_016860030.1
12
1440
XM_017004542.1
XP_016860031.1
11
1350
XM_017004543.1
XP_016860032.1
11
1353
XM_017004544.1
XP_016860033.1
11
1350
XM_017004545.1
XP_016860034.1
14
1841
XM_017004546.1
XP_016860035.1
14
1842
XM_017004547.1
XP_016860036.1
13
1655
XM_017004548.1
XP_016860037.1
13
1629
XM_017004549.1
XP_016860038.1
13
1717
XM_017004550.1
XP_016860039.1
12
1562
XM_017004551.1
XP_016860040.1
8
938
XM_017004552.1
XP_016860041.1
12
1459
XM_017004553.1
XP_016860042.1
12
1459
XM_017004554.1
XP_016860043.1
2
642
GenBank mRNA
AB040457.1
-
11
1418
AK293793.1
-
12
1559
AK294767.1
-
11
1449
AK308529.1
-
15
1932
AY376402.1
-
12
1416
BC136269.1
-
11
1396
9020256
BC143714.1
-
12
1486
9052228
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
chloride transport
nervous system development
response to light stimulus
post-embryonic development
bicarbonate transport
pyramidal neuron development
multicellular organism growth
locomotory exploration behavior
brain morphogenesis
regulation of intracellular pH
anion transmembrane transport
hydrogen ion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
symporter activity
anion:anion antiporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0