Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134771.1 | NP_001128243.1 | 6, 7 | 748 | ||
NM_020708.4 | NP_065759.1 | 6, 7 | 766 | |||
XM_017027981.1 | XP_016883470.1 | 6, 7 | 748 | |||
GenBank mRNA | AB033002.1 | - | 6, 7 | 581 | ||
AF208159.1 | - | 6, 7 | 603 | |||
AK098371.1 | - | 6, 7 | 752 | |||
AK289758.1 | - | 6, 7 | 725 | |||
BC132668.1 | - | 6, 7 | 691 | 40146690 | ||
BC132670.1 | - | 6, 7 | 691 | 40146692 | ||
KC295694.1 | - | 5, 6 | 1127 | |||
KC295701.1 | - | 6, 7 | 766 |