Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020713.2 | NP_065764.1 | 5 | 535 | ||
XM_005260226.3 | XP_005260283.1 | 6 | 607 | |||
XM_011528929.2 | XP_011527231.1 | 5 | 497 | |||
XM_011528930.2 | XP_011527232.1 | 5 | 724 | |||
XM_011528933.2 | XP_011527235.1 | 5 | 720 | |||
GenBank mRNA | AB033022.1 | - | 3 | 354 | ||
BC125128.1 | - | 5 | 526 | 40119047 | ||
BC125129.1 | - | 5 | 525 | 40119053 |