Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 15, 16 | 2602 | ||
XM_005247681.3 | XP_005247738.1 | 9, 10 | 1587 | |||
XM_006713715.1 | XP_006713778.1 | 14, 15 | 2544 | |||
XM_011513051.1 | XP_011511353.1 | 14, 15 | 3051 | |||
XM_011513052.2 | XP_011511354.1 | 13, 14 | 2358 | |||
XM_011513053.1 | XP_011511355.1 | 14, 15 | 2494 | |||
XM_011513054.2 | XP_011511356.1 | 14, 15 | 2330 | |||
XM_017006956.1 | XP_016862445.1 | 13, 14 | 2464 | |||
XM_017006957.1 | XP_016862446.1 | 14, 15 | 3149 | |||
XM_017006958.1 | XP_016862447.1 | 14, 15 | 2996 | |||
XM_017006959.1 | XP_016862448.1 | 12, 13 | 2090 | |||
GenBank mRNA | AB037828.1 | - | 11, 12 | 1880 | ||
AF509494.1 | - | 15, 16 | 2602 | |||
AK302488.1 | - | 15, 16 | 2853 | |||
BC150592.1 | - | 13, 14 | 2602 |