Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UNC79 SIRNA Assays ›
Gene Symbol:
UNC79
Gene Name:
unc-79 homolog (C. elegans)
Gene Aliases:
KIAA1409
Chromosome Location:
Chr.14: 93333219 - 93710473 on Build GRCh38
UniGene ID:
Hs.126561
Species:
Human
siRNA ID
HSS126495
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UNC79
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020818.3
NP_065869.3
30
4902
XM_011537018.2
XP_011535320.1
32
7535
XM_011537020.2
XP_011535322.1
31
7469
XM_011537021.2
XP_011535323.1
32
5086
XM_011537022.2
XP_011535324.1
32
7379
XM_011537023.2
XP_011535325.1
32
7376
XM_011537024.2
XP_011535326.1
31
7313
XM_011537025.2
XP_011535327.1
32
7535
XM_011537026.2
XP_011535328.1
32
8028
XM_011537027.2
XP_011535329.1
32
5053
XM_011537028.2
XP_011535330.1
31
5052
XM_011537029.2
XP_011535331.1
23
4001
XM_017021506.1
XP_016876995.1
32
7433
XM_017021507.1
XP_016876996.1
32
7535
XM_017021508.1
XP_016876997.1
31
7385
XM_017021509.1
XP_016876998.1
29
7376
XM_017021510.1
XP_016876999.1
31
7469
XM_017021511.1
XP_016877000.1
27
4329
XM_017021512.1
XP_016877001.1
31
5020
XM_017021513.1
XP_016877002.1
30
7319
XM_017021514.1
XP_016877003.1
31
7385
XM_017021515.1
XP_016877004.1
32
4934
XM_017021516.1
XP_016877005.1
30
7319
XM_017021517.1
XP_016877006.1
30
7160
XM_017021518.1
XP_016877007.1
26
5457
XM_017021519.1
XP_016877008.1
22
4551
XM_017021520.1
XP_016877009.1
16
2856
GenBank mRNA
AB037830.2
-
28
4256
Back To Top
More Information
Gene Ontology Categories:
Function(s)
ion transmembrane transport
multicellular organism growth
behavioral response to ethanol
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0