Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020830.4 | NP_065881.1 | 8, 9 | 939 | ||
GenBank mRNA | AB037856.1 | - | 8, 9 | 852 | ||
AF411977.1 | - | 8, 9 | 836 | |||
AJ310568.1 | - | 8, 9 | 926 | |||
AK022888.1 | - | 8, 9 | 865 | |||
AK023415.1 | - | 7, 8 | 733 | |||
BC040525.1 | - | 8, 9 | 851 | 5285567 | ||
BC065934.1 | - | 8, 9 | 883 | 6164567 |