Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 7 | 2053 | ||
XM_006712654.2 | XP_006712717.1 | 7 | 2630 | |||
XM_006712655.3 | XP_006712718.1 | 3 | 329 | |||
XM_011511530.2 | XP_011509832.1 | 6 | 1765 | |||
XM_017004569.1 | XP_016860058.1 | 6 | 1757 | |||
XM_017004570.1 | XP_016860059.1 | 6 | 1757 | |||
XM_017004571.1 | XP_016860060.1 | 6 | 1757 | |||
XM_017004572.1 | XP_016860061.1 | 4 | 1242 | |||
GenBank mRNA | AB046783.2 | - | 6 | 1774 | ||
AB053305.1 | - | 7 | 1820 | |||
AF391100.1 | - | 7 | 1820 | |||
AK291029.1 | - | 7 | 1837 | |||
BC029174.1 | - | 7 | 1823 | |||
BC071576.1 | - | 7 | 1815 | 30347073 | ||
KU178735.1 | - | 6 | 1697 | |||
KU178736.1 | - | 6 | 1697 |