Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC46A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033051.3 | NP_149040.3 | 1 | 1000 | ||
GenBank mRNA | AF242557.1 | - | 1 | 1000 | ||
AK055571.1 | - | 2 | 940 | |||
AK314414.1 | - | 1 | 907 | |||
BC048285.1 | - | 1 | 852 | 5213610 |