Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRUNE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303229.1 | NP_001290158.1 | 7 | 1211 | ||
NM_001303242.1 | NP_001290171.1 | 7 | 1255 | |||
NM_001303243.1 | NP_001290172.1 | 6 | 1070 | |||
NM_021222.2 | NP_067045.1 | 8 | 1414 | |||
NR_130130.1 | 4 | 774 | ||||
NR_130131.1 | 5 | 867 | ||||
NR_130132.1 | 4 | 708 | ||||
NR_130135.1 | 5 | 867 | ||||
XM_005245393.4 | XP_005245450.1 | 8 | 1302 | |||
XM_011509832.2 | XP_011508134.1 | 6 | 1103 | |||
XM_017001955.1 | XP_016857444.1 | 7 | 1143 | |||
XM_017001956.1 | XP_016857445.1 | 6 | 1012 | |||
XM_017001957.1 | XP_016857446.1 | 6 | 959 | |||
GenBank mRNA | AF051907.1 | - | 8 | 1328 | ||
AF123538.1 | - | 4 | 688 | |||
AF123539.1 | - | 7 | 1095 | |||
AK027875.1 | - | 4 | 547 | |||
AK294154.1 | - | 5 | 728 | |||
AK298273.1 | - | 4 | 542 | |||
AK315120.1 | - | 8 | 1345 | |||
AK316537.1 | - | 2 | 344 | |||
AL122054.1 | - | 1 | 72 | |||
BC014886.1 | - | 7 | 1119 | 3907577 | ||
BC025304.1 | - | 8 | 1308 | 5458046 | ||
BC063481.1 | - | 4 | 569 | |||
U67085.1 | - | 7 | 1120 |