Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KMT2C SIRNA Assays ›
Gene Symbol:
KMT2C
Gene Name:
lysine methyltransferase 2C
Gene Aliases:
HALR, MLL3
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
UniGene ID:
Hs.647120
Species:
Human
siRNA ID
HSS126898
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_170606.2
NP_733751.2
36, 37
7378
XM_005250025.4
XP_005250082.1
38, 39
7232
XM_005250026.3
XP_005250083.1
38, 39
7266
XM_005250027.4
XP_005250084.1
37, 38
7440
XM_005250028.4
XP_005250085.1
37, 38
7440
XM_005250031.4
XP_005250088.1
37, 38
7440
XM_006716077.3
XP_006716140.1
37, 38
7440
XM_006716078.3
XP_006716141.1
37, 38
7440
XM_006716079.3
XP_006716142.1
37, 38
7440
XM_011516450.2
XP_011514752.1
36, 37
7392
XM_011516451.2
XP_011514753.1
36, 37
7320
XM_011516452.2
XP_011514754.1
35, 36
7287
XM_011516453.2
XP_011514755.1
37, 38
7440
XM_011516454.2
XP_011514756.1
32, 33
6435
XM_011516456.2
XP_011514758.1
36, 37
7392
XM_017012480.1
XP_016867969.1
38, 39
7269
XM_017012481.1
XP_016867970.1
38, 39
7229
XM_017012482.1
XP_016867971.1
37, 38
7440
XM_017012483.1
XP_016867972.1
37, 38
7440
XM_017012484.1
XP_016867973.1
37, 38
7407
XM_017012485.1
XP_016867974.1
36, 37
7389
XM_017012486.1
XP_016867975.1
37, 38
7440
XM_017012487.1
XP_016867976.1
37, 38
7408
XM_017012488.1
XP_016867977.1
36, 37
7257
XM_017012489.1
XP_016867978.1
18, 19
3947
XM_017012490.1
XP_016867979.1
15, 16
3519
GenBank mRNA
AB040939.2
-
25, 26
5494
AF264750.2
-
22, 23
4705
AY024361.1
-
36, 37
7339
BC151837.1
-
25, 26
5494
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0