Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303051.1 | NP_001289980.1 | 12 | 1826 | ||
NM_001303109.1 | NP_001290038.1 | 10 | 1543 | |||
NM_021633.3 | NP_067646.1 | 12 | 1846 | |||
XM_011509835.1 | XP_011508137.1 | 13 | 1866 | |||
XM_011509836.1 | XP_011508138.1 | 12 | 1679 | |||
XM_011509837.1 | XP_011508139.1 | 11 | 1563 | |||
XM_017001995.1 | XP_016857484.1 | 11 | 1628 | |||
GenBank mRNA | AF190900.1 | - | 12 | 1770 | ||
AF306687.1 | - | 8 | 933 | |||
AK024412.1 | - | 6 | 750 | |||
AK027656.1 | - | 12 | 1817 | |||
AK130662.1 | - | 2 | 233 | |||
AK223072.1 | - | 12 | 1782 | |||
AK301791.1 | - | 12 | 1781 | |||
BC003183.1 | - | 12 | 1798 | 2958852 | ||
BC004175.2 | - | 12 | 1798 | 2958852 |