Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021639.4 | NP_067652.1 | 8, 9 | 2111 | ||
XM_017002019.1 | XP_016857508.1 | 8, 9 | 2172 | |||
GenBank mRNA | AF177334.1 | - | 6, 7 | 929 | ||
AK024972.1 | - | 6, 7 | 918 | |||
AK225197.1 | - | 6, 7 | 918 | |||
AL834194.1 | - | 6, 7 | 1761 | |||
BC015713.1 | - | 6, 7 | 1010 | 3909296 | ||
BC022218.1 | - | 6, 7 | 1194 | 3900818 | ||
BC052257.1 | - | 5, 6 | 759 | 6297561 | ||
BC068578.1 | - | 8, 9 | 2091 |