Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286116.1 | NP_001273045.1 | 3 | 483 | ||
NM_001286117.1 | NP_001273046.1 | 3 | 1030 | |||
NM_021825.4 | NP_068597.2 | 3 | 710 | |||
XM_005274159.3 | XP_005274216.1 | 4 | 376 | |||
XM_017018104.1 | XP_016873593.1 | 4 | 801 | |||
GenBank mRNA | AF182412.1 | - | 3 | 280 | ||
AF182424.1 | - | 3 | 280 | |||
AF271782.1 | - | 3 | 637 | |||
AK055972.1 | - | 3 | 1027 | |||
AK225726.1 | - | 3 | 291 | |||
AK304769.1 | - | 3 | 443 | |||
AK315787.1 | - | 3 | 671 | |||
AL136791.1 | - | 3 | 430 | |||
BC014573.1 | - | 3 | 320 | 3680689 | ||
BC020783.1 | - | 3 | 281 | 4773466 | ||
BC067764.1 | - | 3 | 601 | |||
BC107755.1 | - | 3 | 451 | 6737596 | ||
BF131344.1 | - | 376 |