Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021927.2 | NP_068746.2 | 8 | 1129 | ||
XM_005248122.3 | XP_005248179.1 | 7 | 983 | |||
XM_011513732.2 | XP_011512034.1 | 8 | 1150 | |||
GenBank mRNA | AK023282.1 | - | 8 | 1079 | ||
AK225314.1 | - | 8 | 1084 | |||
AK308667.1 | - | 7 | 939 | |||
BC036768.1 | - | 8 | 1090 | 5744283 |