Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021927.2 | NP_068746.2 | 11 | 1468 | ||
XM_005248122.3 | XP_005248179.1 | 10 | 1322 | |||
XM_011513732.2 | XP_011512034.1 | 11 | 1489 | |||
GenBank mRNA | AK023282.1 | - | 11 | 1418 | ||
AK025248.1 | - | 3 | 248 | |||
AK225314.1 | - | 11 | 1423 | |||
BC036768.1 | - | 11 | 1429 | 5744283 |