Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021933.3 | NP_068752.2 | 3 | 569 | ||
XM_005263487.3 | XP_005263544.1 | 3 | 398 | |||
XM_011541895.1 | XP_011540197.1 | 3 | 400 | |||
XM_011541896.1 | XP_011540198.1 | 3 | 293 | |||
GenBank mRNA | AK022500.1 | - | 3 | 349 | ||
AK024020.1 | - | 3 | 288 | |||
BC008068.2 | - | 3 | 278 | 3050071 | ||
BG536799.1 | - | 3 | 357 | |||
BI828925.1 | - | 3 | 326 | |||
FJ618905.1 | - | 3 | 569 |