Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: SRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304803.1 | NP_001291732.1 | 2, 3 | 192 | ||
NM_021947.2 | NP_068766.1 | 3, 4 | 364 | |||
XM_006721565.3 | XP_006721628.1 | 3, 4 | 396 | |||
XM_006721566.3 | XP_006721629.1 | 3, 4 | 1574 | |||
XM_011523974.2 | XP_011522276.1 | 3, 4 | 503 | |||
GenBank mRNA | AF169974.1 | - | 2, 3 | 301 | ||
AK023169.1 | - | 3, 4 | 353 | |||
AK223120.1 | - | 3, 4 | 357 | |||
AL834378.1 | - | 1, 2 | 129 | |||
AY034081.1 | - | 2, 3 | 285 | |||
BC074728.2 | - | 2, 3 | 303 | 30915181 | ||
CR457300.1 | - | 2, 3 | 285 | |||
DA297969.1 | - | 2, 3 | 181 |