Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 15 | 1608 | ||
NM_001193315.1 | NP_001180244.1 | 15 | 1208 | |||
NM_001193316.1 | NP_001180245.1 | 14 | 1061 | |||
NM_001193317.1 | NP_001180246.1 | 15 | 1385 | |||
NM_022067.3 | NP_071350.2 | 16 | 1479 | |||
XM_011537066.1 | XP_011535368.1 | 14 | 1106 | |||
XM_017021579.1 | XP_016877068.1 | 14 | 1240 | |||
XM_017021580.1 | XP_016877069.1 | 15 | 1199 | |||
XM_017021581.1 | XP_016877070.1 | 15 | 1333 | |||
GenBank mRNA | AK022769.1 | - | 15 | 1156 | ||
AK022925.1 | - | 15 | 1134 | |||
AK024661.1 | - | 4 | 304 | |||
AK098254.1 | - | 15 | 1569 | |||
AK222706.1 | - | 15 | 1154 | |||
AK298354.1 | - | 14 | 1007 | |||
BC015054.1 | - | 16 | 1400 | 3920459 | ||
CR457294.1 | - | 14 | 1062 |