Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 4 | 773 | ||
NM_001193315.1 | NP_001180244.1 | 4 | 373 | |||
NM_001193317.1 | NP_001180246.1 | 4 | 550 | |||
NM_022067.3 | NP_071350.2 | 5 | 644 | |||
XM_011537066.1 | XP_011535368.1 | 4 | 364 | |||
XM_017021579.1 | XP_016877068.1 | 4 | 498 | |||
XM_017021580.1 | XP_016877069.1 | 4 | 364 | |||
XM_017021581.1 | XP_016877070.1 | 4 | 498 | |||
GenBank mRNA | AK022769.1 | - | 4 | 321 | ||
AK022925.1 | - | 4 | 299 | |||
AK098254.1 | - | 4 | 734 | |||
AK222706.1 | - | 4 | 319 | |||
AK297388.1 | - | 4 | 475 | |||
BC015054.1 | - | 5 | 565 | 3920459 | ||
CR457294.1 | - | 3 | 227 | |||
DA794284.1 | - | 4 | 373 |