Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174159.1 | NP_001167630.1 | 7 | 1183 | ||
NM_001174160.1 | NP_001167631.1 | 6 | 948 | |||
NM_022071.3 | NP_071354.2 | 7 | 1333 | |||
XM_011544610.1 | XP_011542912.1 | 6 | 1379 | |||
XM_017013729.1 | XP_016869218.1 | 6 | 1101 | |||
GenBank mRNA | AK024799.1 | - | 7 | 1211 | ||
AK293301.1 | - | 6 | 824 | |||
AY190323.1 | - | 3 | 362 | |||
BC014525.2 | - | 4 | 529 | |||
BC067117.1 | - | 4 | 543 | |||
BC082982.1 | - | 7 | 1183 | 6575224 |