Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303472.1 | NP_001290401.1 | 7 | 1347 | ||
NM_022096.5 | NP_071379.3 | 7 | 1365 | |||
NM_198798.2 | NP_942093.1 | 6 | 1301 | |||
XM_005260792.2 | XP_005260849.1 | 5 | 834 | |||
GenBank mRNA | AK097689.1 | - | 6 | 1234 | ||
BC022878.2 | - | 7 | 1275 | 4821619 |