Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022110.3 | NP_071393.2 | 2 | 491 | ||
GenBank mRNA | AK027119.1 | - | 2 | 489 | ||
AK291418.1 | - | 2 | 486 | |||
BC004168.2 | - | 2 | 476 | 2820874 | ||
BC011966.1 | - | 2 | 491 | 3857383 | ||
CR457374.1 | - | 1 | 221 | |||
KP861813.1 | - | 2 | 477 |