Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARVG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137605.2 | NP_001131077.1 | 5 | 655 | ||
NM_022141.6 | NP_071424.1 | 5 | 715 | |||
XM_005261702.3 | XP_005261759.1 | 5 | 442 | |||
XM_011530302.2 | XP_011528604.1 | 4 | 3207 | |||
XM_017028907.1 | XP_016884396.1 | 5 | 1653 | |||
XM_017028908.1 | XP_016884397.1 | 5 | 1673 | |||
GenBank mRNA | AF237772.1 | - | 5 | 509 | ||
AK225929.1 | - | 5 | 658 | |||
AL355092.1 | - | 4 | 422 | |||
AL590887.1 | - | 3 | 231 | |||
BC034406.1 | - | 5 | 636 | 5178529 | ||
BI818662.1 | - | 5 | 462 | |||
BP299720.1 | - | 5 | 481 | |||
BX648942.1 | - | 1 | 2337 | |||
CR456480.2 | - | 4 | 286 |