Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022142.4 | NP_071425.3 | 4 | 404 | ||
XM_017027130.1 | XP_016882619.1 | 4 | 503 | |||
GenBank mRNA | AF279147.1 | - | 4 | 404 | ||
AJ278478.1 | - | 3 | 245 | |||
BC015598.2 | - | 4 | 368 | 4649702 | ||
GQ149084.1 | - | 4 | 368 |