Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324159.1 | NP_001311088.1 | 3 | 282 | ||
NM_022355.3 | NP_071750.1 | 5 | 775 | |||
NR_136706.1 | 6 | 1252 | ||||
XM_005256090.4 | XP_005256147.2 | 4 | 1732 | |||
XM_011523266.1 | XP_011521568.1 | 5 | 718 | |||
XM_011523268.2 | XP_011521570.1 | 4 | 1732 | |||
XM_011523271.2 | XP_011521573.1 | 4 | 1732 | |||
XM_011523272.2 | XP_011521574.1 | 4 | 689 | |||
XM_011523273.2 | XP_011521575.1 | 3 | 381 | |||
XM_017023541.1 | XP_016879030.1 | 3 | 1381 | |||
XM_017023543.1 | XP_016879032.1 | 4 | 709 | |||
XM_017023544.1 | XP_016879033.1 | 4 | 688 | |||
XM_017023545.1 | XP_016879034.1 | 3 | 966 | |||
XM_017023546.1 | XP_016879035.1 | 3 | 1149 | |||
GenBank mRNA | AJ295149.1 | - | 5 | 710 | ||
AK092884.1 | - | 6 | 1252 | |||
AK298003.1 | - | 3 | 722 | |||
AK315090.1 | - | 4 | 752 | |||
AY358454.1 | - | 4 | 536 |