Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022451.10 | NP_071896.8 | 21 | 2507 | ||
XM_017016544.1 | XP_016872033.1 | 19 | 2560 | |||
GenBank mRNA | AB077992.1 | - | 21 | 2394 | ||
AK022882.1 | - | 13 | 1418 | |||
AK026953.1 | - | 11 | 1118 | |||
AK091246.1 | - | 21 | 2460 | |||
AK292520.1 | - | 21 | 2489 | |||
BC049850.1 | - | 21 | 2457 | 5549517 |