Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006849.2 | NP_006840.2 | 7, 8 | 1165 | ||
GenBank mRNA | AK075485.1 | - | 8, 9 | 1082 | ||
AK124931.1 | - | 6, 7 | 1184 | |||
AK295383.1 | - | 7, 8 | 1057 | |||
AK308516.1 | - | 6, 7 | 1113 | |||
BC000537.2 | - | 7, 8 | 1099 | |||
BC075029.2 | - | 7, 8 | 1097 | |||
U19948.1 | - | 7, 8 | 1098 | |||
Z84717.1 | - | 3, 4 | 1393 |