Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042574.2 | NP_001036039.1 | 5, 6 | 672 | ||
NM_022769.4 | NP_073606.3 | 5, 6 | 672 | |||
XM_005254968.2 | XP_005255025.1 | 5, 6 | 643 | |||
XM_011521906.2 | XP_011520208.1 | 4, 5 | 544 | |||
GenBank mRNA | AK074170.1 | - | 2, 3 | 143 | ||
AK090443.1 | - | 5, 6 | 632 | |||
AY360173.1 | - | 5, 6 | 493 | |||
BQ945777.1 | - | 5, 6 | 621 |