Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022774.1 | NP_073611.1 | 3 | 429 | ||
XM_005271125.3 | XP_005271182.1 | 4 | 677 | |||
XM_005271126.3 | XP_005271183.1 | 3 | 489 | |||
XM_005271128.3 | XP_005271185.1 | 2 | 426 | |||
XM_011541970.2 | XP_011540272.1 | 2 | 371 | |||
XM_017002099.1 | XP_016857588.1 | 4 | 1101 | |||
XM_017002100.1 | XP_016857589.1 | 4 | 656 | |||
XM_017002101.1 | XP_016857590.1 | 4 | 627 | |||
XM_017002102.1 | XP_016857591.1 | 3 | 1029 | |||
XM_017002103.1 | XP_016857592.1 | 3 | 455 | |||
XM_017002104.1 | XP_016857593.1 | 2 | 408 | |||
GenBank mRNA | AK023245.1 | - | 2 | 1256 | ||
AK024797.1 | - | 3 | 429 | |||
AK094092.1 | - | 3 | 1251 | |||
BC021969.2 | - | 4 | 590 | 3952573 |