Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163315.2 | NP_001156787.2 | 8, 9 | 2363 | ||
GenBank mRNA | AB533280.1 | - | 9, 10 | 1283 | ||
AK126722.1 | - | 9, 10 | 1551 | |||
BC126144.1 | - | 8, 9 | 1089 | 8991860 | ||
BC126146.1 | - | 8, 9 | 1089 | 8991862 |