Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022841.5 | NP_073752.5 | 9 | 1754 | ||
XM_005254603.3 | XP_005254660.2 | 10 | 1848 | |||
XM_011521925.2 | XP_011520227.1 | 9 | 1686 | |||
XM_017022506.1 | XP_016877995.1 | 10 | 4571 | |||
XM_017022507.1 | XP_016877996.1 | 9 | 1814 | |||
XM_017022508.1 | XP_016877997.1 | 8 | 2983 | |||
GenBank mRNA | AK023056.1 | - | 1 | 49 | ||
AK126103.1 | - | 2 | 906 | |||
AK128045.1 | - | 9 | 1691 | |||
AL832584.1 | - | 1 | 564 | |||
BC112936.1 | - | 7 | 1574 | 6014850 | ||
BC143438.1 | - | 7 | 1511 |