Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 11 | 1067 | ||
NM_001321996.1 | NP_001308925.1 | 11 | 1167 | |||
NM_001321997.1 | NP_001308926.1 | 11 | 988 | |||
NM_031902.4 | NP_114108.1 | 11 | 1246 | |||
NR_136152.1 | 10 | 1076 | ||||
NR_136153.1 | 10 | 1155 | ||||
GenBank mRNA | AB049940.1 | - | 11 | 1127 | ||
AK055390.1 | - | 1 | 1860 | |||
AK058160.1 | - | 10 | 929 | |||
AK124892.1 | - | 1 | 3248 | |||
BC000219.2 | - | 7 | 561 | |||
BC014172.2 | - | 11 | 1219 | 4561399 |