Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 3 | 309 | ||
NM_001321996.1 | NP_001308925.1 | 3 | 488 | |||
NM_001321997.1 | NP_001308926.1 | 3 | 309 | |||
NM_031902.4 | NP_114108.1 | 3 | 488 | |||
NR_136152.1 | 3 | 488 | ||||
NR_136153.1 | 3 | 488 | ||||
GenBank mRNA | AB049940.1 | - | 3 | 369 | ||
AK058160.1 | - | 3 | 262 | |||
AK295812.1 | - | 3 | 274 | |||
BC014172.2 | - | 3 | 461 | 4561399 | ||
BG771419.1 | - | 3 | 475 |