Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MPPE1 SIRNA Assays ›
Gene Symbol:
MPPE1
Gene Name:
metallophosphoesterase 1
Gene Aliases:
PGAP5
Chromosome Location:
Chr.18: 11883471 - 11908797 on Build GRCh38
UniGene ID:
Hs.712666
Species:
Human
siRNA ID
HSS128022
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MPPE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242904.1
NP_001229833.1
4, 5
1180
NM_001319154.1
NP_001306083.1
5, 6
1287
NM_023075.5
NP_075563.3
5, 6
1287
NR_040241.1
5, 6
1287
NR_040242.3
6, 7
1155
NR_040243.1
5, 6
1287
XM_006722340.3
XP_006722403.1
5, 6
840
XM_006722341.3
XP_006722404.1
7, 8
1462
XM_006722342.3
XP_006722405.1
6, 7
1246
XM_006722343.3
XP_006722406.1
6, 7
1367
XM_006722345.3
XP_006722408.1
4, 5
1180
XM_006722346.2
XP_006722409.1
4, 5
1180
XM_006722347.3
XP_006722410.1
5, 6
840
XM_006722348.2
XP_006722411.1
5, 6
840
XM_011525731.2
XP_011524033.1
8, 9
1870
XM_011525732.1
XP_011524034.1
5, 6
840
XM_011525734.1
XP_011524036.1
5, 6
840
XM_017025920.1
XP_016881409.1
7, 8
1775
XM_017025921.1
XP_016881410.1
4, 5
682
XM_017025922.1
XP_016881411.1
7, 8
1462
XM_017025923.1
XP_016881412.1
8, 9
1870
XM_017025924.1
XP_016881413.1
6, 7
1246
XM_017025925.1
XP_016881414.1
6, 7
1367
XM_017025926.1
XP_016881415.1
7, 8
1775
XM_017025927.1
XP_016881416.1
6, 7
1246
XM_017025928.1
XP_016881417.1
6, 7
1367
XM_017025929.1
XP_016881418.1
4, 5
1180
XM_017025930.1
XP_016881419.1
4, 5
1180
XM_017025931.1
XP_016881420.1
5, 6
840
GenBank mRNA
AF289560.1
-
5, 6
808
AF363484.1
-
5, 6
1289
AK023052.1
-
3, 4
504
AK054639.1
-
6, 7
653
AK223450.1
-
4, 5
660
AK309948.1
-
5, 6
805
AL833636.1
-
5, 6
839
AX775853.1
-
5, 6
837
BC002877.2
-
4, 5
667
3942160
BC073994.1
-
5, 6
799
6180123
Back To Top
More Information
Panther Classification:
Molecular Function -
esterase
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
ER to Golgi vesicle-mediated transport
protein binding
phosphoric diester hydrolase activity
manganese ion binding
GPI anchor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0