Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK33 SIRNA Assays ›
Gene Symbol:
STK33
Gene Name:
serine/threonine kinase 33
Gene Aliases:
-
Chromosome Location:
Chr.11: 8335029 - 8594289 on Build GRCh38
UniGene ID:
Hs.501833
Species:
Human
siRNA ID
HSS128041
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STK33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289058.1
NP_001275987.1
13
1465
NM_001289059.1
NP_001275988.1
10
1172
NM_001289061.1
NP_001275990.1
12
1730
NM_030906.3
NP_112168.1
13
1879
XM_011520288.2
XP_011518590.1
13
2957
XM_011520290.2
XP_011518592.1
15
1876
XM_011520292.2
XP_011518594.1
13
1678
XM_011520293.2
XP_011518595.1
13
1707
XM_011520294.1
XP_011518596.1
12
1672
XM_011520295.2
XP_011518597.1
12
3768
XM_011520297.2
XP_011518599.1
12
1445
XM_017018143.1
XP_016873632.1
15
1936
XM_017018144.1
XP_016873633.1
15
1838
XM_017018145.1
XP_016873634.1
14
3870
XM_017018146.1
XP_016873635.1
15
2000
XM_017018147.1
XP_016873636.1
14
3797
XM_017018148.1
XP_016873637.1
13
3729
XM_017018149.1
XP_016873638.1
14
1881
XM_017018150.1
XP_016873639.1
14
1736
XM_017018151.1
XP_016873640.1
14
1695
XM_017018152.1
XP_016873641.1
12
2026
XM_017018153.1
XP_016873642.1
12
1624
XM_017018154.1
XP_016873643.1
12
1483
XM_017018155.1
XP_016873644.1
12
2038
XM_017018156.1
XP_016873645.1
12
1484
XM_017018157.1
XP_016873646.1
12
1249
XM_017018158.1
XP_016873647.1
13
1535
XM_017018165.1
XP_016873654.1
13
1438
GenBank mRNA
AJ303380.2
-
11
2223
AK092720.1
-
10
1111
AK093251.1
-
12
1588
AK293193.1
-
13
1331
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein autophosphorylation
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0