Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD3 SIRNA Assays ›
Gene Symbol:
GRAMD3
Gene Name:
GRAM domain containing 3
Gene Aliases:
NS3TP2
Chromosome Location:
Chr.5: 126360096 - 126494377 on Build GRCh38
UniGene ID:
Hs.363558
Species:
Human
siRNA ID
HSS128060
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146319.1
NP_001139791.1
13
1472
NM_001146320.1
NP_001139792.1
12
1509
NM_001146321.1
NP_001139793.1
14
1664
NM_001146322.1
NP_001139794.1
13
1283
NM_023927.2
NP_076416.2
13
1621
XM_005272057.3
XP_005272114.1
13
1362
XM_005272058.3
XP_005272115.1
13
1359
XM_005272059.4
XP_005272116.1
14
1752
XM_005272060.3
XP_005272117.1
13
1449
XM_005272061.3
XP_005272118.1
13
1444
XM_005272062.3
XP_005272119.1
13
1410
XM_005272063.3
XP_005272120.1
13
1556
XM_005272064.3
XP_005272121.1
14
1469
XM_011543591.2
XP_011541893.1
14
2340
XM_011543592.2
XP_011541894.1
14
2333
XM_011543593.2
XP_011541895.1
13
1449
XM_011543594.2
XP_011541896.1
13
1221
XM_011543595.2
XP_011541897.1
12
1216
XM_017009781.1
XP_016865270.1
14
1227
XM_017009782.1
XP_016865271.1
14
1466
XM_017009783.1
XP_016865272.1
12
1077
XM_017009784.1
XP_016865273.1
12
1268
XM_017009785.1
XP_016865274.1
12
1318
GenBank mRNA
AK024966.1
-
13
1389
AK225829.1
-
13
1389
AK293307.1
-
12
1214
AK294656.1
-
13
1270
AK296267.1
-
14
1369
AK298009.1
-
13
1206
AK298349.1
-
13
1149
AK300154.1
-
13
1417
AK303857.1
-
13
1223
AK316288.1
-
13
1436
AY116970.1
-
13
1191
BC008590.1
-
13
1354
4148772
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0