Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300883.2 | NP_001287812.1 | 6 | 1710 | ||
NM_001318055.1 | NP_001304984.1 | 7 | 1446 | |||
NM_001318056.1 | NP_001304985.1 | 7 | 1490 | |||
NM_024106.2 | NP_077011.1 | 8 | 1573 | |||
XM_017027293.1 | XP_016882782.1 | 7 | 1509 | |||
XM_017027294.1 | XP_016882783.1 | 5 | 1615 | |||
GenBank mRNA | AB209837.1 | - | 6 | 1655 | ||
AK095759.1 | - | 6 | 1628 | |||
BC001791.1 | - | 8 | 1555 | 3543910 |