Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024512.4 | NP_078788.2 | 4 | 865 | ||
XM_011534110.2 | XP_011532412.1 | 4 | 654 | |||
XM_017007177.1 | XP_016862666.1 | 4 | 865 | |||
GenBank mRNA | AJ308569.2 | - | 4 | 639 | ||
AK057808.1 | - | 4 | 707 | |||
AK223440.1 | - | 4 | 707 | |||
AK314658.1 | - | 4 | 628 | |||
AK315636.1 | - | 4 | 648 | |||
BC029118.1 | - | 4 | 637 | 5183639 |