Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122835.2 | NP_001116307.2 | 18, 19 | 2180 | ||
NM_024525.4 | NP_078801.3 | 20, 21 | 2342 | |||
XM_005273260.1 | XP_005273317.1 | 20, 21 | 2339 | |||
XM_005273261.2 | XP_005273318.1 | 19, 20 | 2249 | |||
XM_005273262.1 | XP_005273319.1 | 18, 19 | 2183 | |||
XM_005273264.2 | XP_005273321.1 | 18, 19 | 2114 | |||
XM_006711814.2 | XP_006711877.1 | 16, 17 | 1955 | |||
XM_006711815.2 | XP_006711878.1 | 13, 14 | 3349 | |||
XM_011544275.2 | XP_011542577.1 | 17, 18 | 2021 | |||
XM_011544276.2 | XP_011542578.1 | 13, 14 | 1585 | |||
XM_017002321.1 | XP_016857810.1 | 19, 20 | 2246 | |||
XM_017002322.1 | XP_016857811.1 | 18, 19 | 2111 | |||
XM_017002323.1 | XP_016857812.1 | 16, 17 | 1917 | |||
XM_017002324.1 | XP_016857813.1 | 16, 17 | 1952 | |||
XM_017002325.1 | XP_016857814.1 | 20, 21 | 5544 | |||
XM_017002326.1 | XP_016857815.1 | 15, 16 | 4924 | |||
GenBank mRNA | AK026237.1 | - | 7, 8 | 649 | ||
AK074365.1 | - | 11, 12 | 1250 | |||
AK075386.1 | - | 20, 21 | 2308 | |||
AK075561.1 | - | 18, 19 | 2159 | |||
AK309725.1 | - | 16, 17 | 1998 | |||
AL832286.1 | - | 20, 21 | 2077 | |||
AL834453.1 | - | 11, 12 | 1308 | |||
BC042683.1 | - | 11, 12 | 1226 |