Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPAG16 SIRNA Assays ›
Gene Symbol:
SPAG16
Gene Name:
sperm associated antigen 16
Gene Aliases:
PF20, WDR29
Chromosome Location:
Chr.2: 213284369 - 214410501 on Build GRCh38
UniGene ID:
Hs.743365
Species:
Human
siRNA ID
HSS128456
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPAG16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025436.2
NP_001020607.1
4, 5
513
NM_024532.4
NP_078808.3
4, 5
513
NR_047659.1
6, 7
688
NR_047660.1
3, 4
394
XM_006712746.2
XP_006712809.1
4, 5
464
XM_011511814.1
XP_011510116.1
3, 4
561
XM_011511815.1
XP_011510117.1
4, 5
464
XM_011511816.2
XP_011510118.1
4, 5
464
XM_011511817.1
XP_011510119.1
4, 5
464
XM_011511818.1
XP_011510120.1
4, 5
464
XM_011511819.2
XP_011510121.1
3, 4
1212
XM_011511820.1
XP_011510122.1
4, 5
464
XM_011511821.2
XP_011510123.1
3, 4
1338
XM_011511822.1
XP_011510124.1
2, 3
194
XM_011511823.2
XP_011510125.1
2, 3
1116
XM_011511824.1
XP_011510126.1
4, 5
464
XM_011511827.1
XP_011510129.1
4, 5
464
XM_011511828.1
XP_011510130.1
4, 5
464
XM_011511829.1
XP_011510131.1
4, 5
464
XM_011511831.1
XP_011510133.1
4, 5
464
XM_011511832.1
XP_011510134.1
4, 5
464
XM_011511835.2
XP_011510137.1
3, 4
556
XM_011511837.2
XP_011510139.1
4, 5
464
XM_017004896.1
XP_016860385.1
5, 6
593
XM_017004897.1
XP_016860386.1
2, 3
3210
XM_017004899.1
XP_016860388.1
4, 5
464
XM_017004900.1
XP_016860389.1
4, 5
455
XM_017004901.1
XP_016860390.1
4, 5
445
GenBank mRNA
AF310672.1
-
4, 5
450
AF426740.1
-
4, 5
450
AF490390.1
-
4, 5
443
AK026377.1
-
4, 5
473
AK095036.1
-
3, 4
561
AK302351.1
-
3, 4
336
AK310883.1
-
4, 5
412
AL832962.1
-
4, 5
402
AW207406.1
-
1
94
BC009614.1
-
4, 5
474
BC025379.1
-
6, 7
693
4820489
BC067756.1
-
4, 5
500
BC100282.1
-
4, 5
461
CR457359.1
-
4, 5
408
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sperm axoneme assembly
axoneme assembly
cilium assembly
microtubule sliding
cilium movement involved in cell motility
cell motility in response to calcium ion
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-462rt:80/10.239.167.23:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0