Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
siRNA ID
HSS128712
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
6
753
NM_001164629.3
NP_001158101.1
6
738
NM_001284233.1
NP_001271162.1
4
562
NM_001284234.1
NP_001271163.1
4
453
NM_001284235.1
NP_001271164.1
6
720
NM_001284238.1
NP_001271167.1
8
952
NM_024659.4
NP_078935.2
6
720
XM_005263774.2
XP_005263831.1
6
690
XM_005263782.2
XP_005263839.1
6
706
XM_005263784.2
XP_005263841.1
4
565
XM_011511843.2
XP_011510145.1
6
739
XM_011511855.2
XP_011510157.1
4
683
XM_011511856.2
XP_011510158.1
4
465
XM_017004917.1
XP_016860406.1
10
1361
XM_017004918.1
XP_016860407.1
8
953
XM_017004919.1
XP_016860408.1
6
839
XM_017004920.1
XP_016860409.1
8
3995
XM_017004921.1
XP_016860410.1
7
788
XM_017004922.1
XP_016860411.1
7
965
XM_017004923.1
XP_016860412.1
6
753
XM_017004924.1
XP_016860413.1
8
949
XM_017004925.1
XP_016860414.1
5
611
XM_017004926.1
XP_016860415.1
4
689
XM_017004927.1
XP_016860416.1
6
720
XM_017004928.1
XP_016860417.1
8
953
XM_017004929.1
XP_016860418.1
6
690
XM_017004930.1
XP_016860419.1
6
1341
XM_017004931.1
XP_016860420.1
6
754
XM_017004932.1
XP_016860421.1
6
724
XM_017004933.1
XP_016860422.1
6
706
XM_017004934.1
XP_016860423.1
7
3913
XM_017004935.1
XP_016860424.1
8
1783
XM_017004936.1
XP_016860425.1
7
960
XM_017004937.1
XP_016860426.1
4
694
XM_017004938.1
XP_016860427.1
4
692
XM_017004939.1
XP_016860428.1
6
742
XM_017004940.1
XP_016860429.1
4
690
XM_017004941.1
XP_016860430.1
4
689
XM_017004942.1
XP_016860431.1
6
736
XM_017004943.1
XP_016860432.1
6
1402
XM_017004944.1
XP_016860433.1
6
735
XM_017004945.1
XP_016860434.1
8
953
XM_017004946.1
XP_016860435.1
6
690
XM_017004947.1
XP_016860436.1
6
1373
XM_017004948.1
XP_016860437.1
7
3913
XM_017004949.1
XP_016860438.1
4
688
XM_017004950.1
XP_016860439.1
6
735
XM_017004951.1
XP_016860440.1
6
690
XM_017004952.1
XP_016860441.1
6
1134
XM_017004953.1
XP_016860442.1
6
706
XM_017004954.1
XP_016860443.1
7
794
XM_017004955.1
XP_016860444.1
9
1142
XM_017004956.1
XP_016860445.1
4
690
XM_017004957.1
XP_016860446.1
6
736
GenBank mRNA
AB201269.1
-
3
474
AB597184.1
-
3
474
AK091684.1
-
4
566
AK291357.1
-
6
668
AY281366.1
-
6
753
BC017741.2
-
7
760
4390876
BC061699.1
-
6
660
4608518
BX647436.1
-
4
435
BX647773.1
-
8
891
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0