Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024712.3 | NP_078988.2 | 6, 7 | 712 | ||
GenBank mRNA | AK023886.1 | - | 5, 6 | 476 | ||
AK056923.1 | - | 5, 6 | 675 | |||
AK300976.1 | - | 6, 7 | 726 | |||
BC034410.1 | - | 3, 4 | 324 |