Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CARF SIRNA Assays ›
Gene Symbol:
CARF
Gene Name:
calcium responsive transcription factor
Gene Aliases:
ALS2CR8, NYD-SP24
Chromosome Location:
Chr.2: 202912218 - 202987856 on Build GRCh38
UniGene ID:
Hs.444982
Species:
Human
siRNA ID
HSS128878
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CARF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104586.2
NP_001098056.1
7
934
NM_001282910.1
NP_001269839.1
6
706
NM_001282911.1
NP_001269840.1
7
787
NM_001282912.1
NP_001269841.1
5
585
NM_001322427.1
NP_001309356.1
8
993
NM_001322428.1
NP_001309357.1
7
1207
NM_001322429.1
NP_001309358.1
6
813
NM_001322430.1
NP_001309359.1
7
934
NM_024744.15
NP_079020.13
8
1831
NR_136330.1
7
934
NR_136331.1
7
934
XM_005246858.3
XP_005246915.1
8
940
XM_005246859.3
XP_005246916.1
8
992
XM_005246864.3
XP_005246921.1
1
56
XM_006712760.2
XP_006712823.1
7
875
XM_011511867.2
XP_011510169.1
8
971
XM_011511868.2
XP_011510170.1
7
875
XM_017004961.1
XP_016860450.1
7
1329
XM_017004962.1
XP_016860451.1
5
561
XM_017004963.1
XP_016860452.1
6
629
XM_017004964.1
XP_016860453.1
5
539
XM_017004965.1
XP_016860454.1
4
347
XM_017004966.1
XP_016860455.1
4
424
GenBank mRNA
AB053309.1
-
8
1314
AB053310.1
-
6
633
AF454946.1
-
5
661
AK025232.1
-
4
534
AK225218.1
-
4
534
AK295499.1
-
5
520
AK295703.1
-
6
745
AK304014.1
-
6
641
AK308171.1
-
7
874
AK310640.1
-
6
754
AK316432.1
-
7
722
AL834188.1
-
7
897
AY032876.2
-
7
886
BC033777.2
-
7
947
4521845
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
cellular response to potassium ion
positive regulation of transcription from RNA polymerase II promoter in response to calcium ion
cellular response to calcium ion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0